siRNA Details
| Gene Symbol: Sorbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205219.1 | NP_001192148.1 | 16 | 2589 | ||
| NM_172752.4 | NP_766340.4 | 15 | 2532 | |||
| GenBank mRNA | AK122369.1 | - | 11 | 2000 | ||
| BC151038.1 | - | 16 | 2459 | 9087951 | ||
| BC172140.1 | - | 15 | 2402 | 9054810 | ||

Sorbs2
Genomic Map