siRNA Details
| Gene Symbol: Helt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173789.4 | NP_776150.2 | 4 | 948 | ||
| GenBank mRNA | AB098077.1 | - | 4 | 399 | ||
| AB120968.1 | - | 4 | 948 | |||
| AK044070.1 | - | 3 | 499 | |||
| BC103599.1 | - | 4 | 833 | 40045925 | ||
| BC103600.1 | - | 4 | 833 | 40045926 | ||
| BC103601.1 | - | 4 | 833 | 40045923 | ||
| DQ294234.1 | - | 4 | 411 | |||

Helt
Genomic Map