siRNA Details
Gene Symbol: BC030500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173411.2 | NP_775587.1 | 2, 3 | 168 | ||
GenBank mRNA | BC030500.1 | - | 2, 3 | 179 | 5401612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC030500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173411.2 | NP_775587.1 | 2, 3 | 168 | ||
GenBank mRNA | BC030500.1 | - | 2, 3 | 179 | 5401612 |