siRNA Details
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252623.1 | NP_001239552.1 | 5 | 1517 | ||
NM_172753.5 | NP_766341.4 | 4 | 1213 | |||
GenBank mRNA | AK028688.1 | - | 4 | 1136 | ||
AK033522.1 | - | 2 | 867 | |||
AK049770.1 | - | 5 | 1811 | |||
AK080687.1 | - | 4 | 1182 | |||
AK133567.1 | - | 2 | 866 | |||
AK144089.1 | - | 1 | 774 | |||
BC057630.1 | - | 6 | 1485 | |||
BC089162.1 | - | 4 | 1210 | 30470205 |
