siRNA Details
Gene Symbol: Tmem161a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145597.4 | NP_663572.1 | 2, 3 | 135 | ||
GenBank mRNA | AK049979.1 | - | 2, 3 | 134 | ||
AK082758.1 | - | 2, 3 | 131 | |||
AK147962.1 | - | 2, 3 | 128 | |||
BC021367.1 | - | 2, 3 | 135 | 5030434 |
