siRNA Details
| Gene Symbol: Heatr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172757.3 | NP_766345.3 | 10 | 1454 | ||
| GenBank mRNA | AK047782.1 | - | 10 | 1485 | ||
| AK052635.1 | - | 10 | 1481 | |||
| AK134510.1 | - | 10 | 1459 | |||
| AK136424.1 | - | 10 | 1464 | |||
| AK140522.1 | - | 10 | 1472 | |||
| AK163715.1 | - | 10 | 2217 | |||
| AK165541.1 | - | 10 | 1348 | |||
| BC030373.1 | - | 6 | 799 | |||
| BC065060.1 | - | 10 | 1454 | 6402217 | ||

Heatr3
Genomic Map