siRNA Details
Gene Symbol: Cpne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153507.2 | NP_705727.1 | 10, 11 | 1126 | ||
GenBank mRNA | BC022128.1 | - | 10, 11 | 1087 | 5097527 | |
BC023348.1 | - | 5, 6 | 424 | 5012573 | ||
BC031801.1 | - | 10, 11 | 1126 | 3985959 |
