siRNA Details
| Gene Symbol: Slc38a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172758.4 | NP_766346.1 | 8 | 1105 | ||
| GenBank mRNA | AK052548.1 | - | 8 | 1106 | ||
| AK171137.1 | - | 8 | 1113 | |||
| AK194183.1 | - | 3 | 221 | |||
| BC031853.1 | - | 6 | 980 | 4009949 | ||

Slc38a7
Genomic Map