siRNA Details
| Gene Symbol: D230025D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145604.2 | NP_663579.2 | 9 | 941 | ||
| GenBank mRNA | AK051965.1 | - | 8 | 829 | ||
| AK156120.1 | - | 9 | 943 | |||
| AK166937.1 | - | 8 | 2128 | |||
| BC006874.1 | - | 9 | 824 | 2646515 | ||
| BC057919.1 | - | 9 | 930 | 5134302 | ||
| DV649781.1 | - | 3 | 194 | |||

D230025D16Rik
Genomic Map