siRNA Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172760.3 | NP_766348.3 | 8, 9 | 892 | ||
| GenBank mRNA | AK037067.1 | - | 8, 9 | 885 | ||
| AK157875.1 | - | 830 | ||||
| BC018516.1 | - | 6, 7 | 781 | 3155913 | ||
| BC026617.1 | - | 3, 4 | 837 | |||
| BC058752.1 | - | 8, 9 | 875 | 6485393 | ||
| BC138887.1 | - | 8, 9 | 849 | 8861909 | ||

Elmo3
Genomic Map