siRNA Details
| Gene Symbol: Fuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172283.3 | NP_758487.2 | 18 | 2283 | ||
| GenBank mRNA | AJ297482.4 | - | 18 | 2254 | ||
| AJ534942.1 | - | 18 | 2254 | |||
| AK049153.1 | - | 7 | 1112 | |||
| AK131165.1 | - | 7 | 3633 | |||
| AK161763.1 | - | 18 | 2387 | |||
| AK164340.1 | - | 18 | 2350 | |||
| AY223865.1 | - | 18 | 2254 | |||
| BC037698.1 | - | 17 | 2232 | 4190449 | ||

Fuk
Genomic Map