siRNA Details
| Gene Symbol: Rfwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146218.4 | NP_666330.2 | 5 | 899 | ||
| GenBank mRNA | AK137499.1 | - | 5 | 879 | ||
| AK137698.1 | - | 5 | 862 | |||
| AK139032.1 | - | 5 | 880 | |||
| AK146451.1 | - | 5 | 869 | |||
| BC018533.1 | - | 5 | 868 | 2651526 | ||
| BC023694.1 | - | 5 | 904 | 5344528 | ||
| BC096602.1 | - | 5 | 862 | 4954068 | ||

Rfwd3
Genomic Map