siRNA Details
| Gene Symbol: Rfwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146218.4 | NP_666330.2 | 10 | 1730 | ||
| GenBank mRNA | AK137499.1 | - | 10 | 1710 | ||
| AK137698.1 | - | 10 | 1693 | |||
| AK139032.1 | - | 10 | 1712 | |||
| BC018533.1 | - | 10 | 1699 | 2651526 | ||
| BC023694.1 | - | 10 | 1735 | 5344528 | ||
| BC027246.1 | - | 5 | 681 | 3497792 | ||
| BC096602.1 | - | 10 | 1693 | 4954068 | ||

Rfwd3
Genomic Map