siRNA Details
| Gene Symbol: Rfwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146218.4 | NP_666330.2 | 13 | 2293 | ||
| GenBank mRNA | AK137499.1 | - | 13 | 2273 | ||
| AK137698.1 | - | 13 | 2256 | |||
| AK139032.1 | - | 13 | 2275 | |||
| BC023694.1 | - | 13 | 2298 | 5344528 | ||
| BC026603.1 | - | 4 | 523 | |||
| BC027246.1 | - | 8 | 1244 | 3497792 | ||
| BC096602.1 | - | 13 | 2256 | 4954068 | ||

Rfwd3
Genomic Map