siRNA Details
Gene Symbol: Nup133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172288.2 | NP_758492.2 | 18 | 2524 | ||
GenBank mRNA | AK029317.1 | - | 18 | 2508 | ||
BC023915.1 | - | 19 | 2535 | 5323157 | ||
BC026845.1 | - | 18 | 2507 | 4235408 | ||
BC033476.1 | - | 18 | 2519 |
