siRNA Details
| Gene Symbol: Nup133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172288.2 | NP_758492.2 | 22, 23 | 3178 | ||
| GenBank mRNA | AK029317.1 | - | 22, 23 | 3162 | ||
| BC023915.1 | - | 23, 24 | 3189 | 5323157 | ||
| BC026845.1 | - | 22, 23 | 3161 | 4235408 | ||
| BC033476.1 | - | 22, 23 | 3173 | |||
| BC058707.1 | - | 3, 4 | 1967 | |||

Nup133
Genomic Map