siRNA Details
| Gene Symbol: Ttc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145607.3 | NP_663582.2 | 7 | 801 | ||
| GenBank mRNA | AK144842.1 | - | 7 | 802 | ||
| AK148102.1 | - | 9 | 1028 | |||
| AK160894.1 | - | 9 | 1039 | |||
| AK183810.1 | - | 2 | 152 | |||
| AK194822.1 | - | 2 | 152 | |||
| BC017545.1 | - | 1 | 15 | |||
| BC072577.1 | - | 7 | 794 | |||
| BC079851.1 | - | 8 | 856 | 6401930 | ||

Ttc13
Genomic Map