siRNA Details
| Gene Symbol: BC021891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145608.2 | NP_663583.2 | 6, 7 | 1854 | ||
| GenBank mRNA | AK220424.1 | - | 6, 7 | 1854 | ||
| BC021891.1 | - | 6, 7 | 1847 | 3156324 | ||
| BC046448.1 | - | 3, 4 | 422 | |||

BC021891
Genomic Map