siRNA Details
Gene Symbol: Med17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144933.1 | NP_659182.1 | 10, 11 | 1644 | ||
GenBank mRNA | AK033151.1 | - | 10, 11 | 1650 | ||
AK038451.1 | - | 10, 11 | 1684 | |||
AK049112.1 | - | 10, 11 | 1684 | |||
AK142033.1 | - | 10, 11 | 1683 | |||
AK151377.1 | - | 10, 11 | 1684 | |||
AK178010.1 | - | 3, 4 | 332 | |||
BC004062.1 | - | 9, 10 | 1321 | |||
BC020185.1 | - | 10, 11 | 1644 | 4505959 |
