siRNA Details
| Gene Symbol: Ccdc67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181816.2 | NP_861537.2 | 5 | 526 | ||
| GenBank mRNA | AK030153.1 | - | 5 | 509 | ||
| BC096547.1 | - | 4 | 419 | 30360184 | ||
| KC211186.1 | - | 4 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc67| Gene Symbol: Ccdc67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181816.2 | NP_861537.2 | 5 | 526 | ||
| GenBank mRNA | AK030153.1 | - | 5 | 509 | ||
| BC096547.1 | - | 4 | 419 | 30360184 | ||
| KC211186.1 | - | 4 | 370 | |||