siRNA Details
| Gene Symbol: Ccdc67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181816.2 | NP_861537.2 | 12 | 1517 | ||
| GenBank mRNA | AK077147.1 | - | 7 | 895 | ||
| BC096547.1 | - | 11 | 1410 | 30360184 | ||
| KC211186.1 | - | 12 | 1448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc67| Gene Symbol: Ccdc67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181816.2 | NP_861537.2 | 12 | 1517 | ||
| GenBank mRNA | AK077147.1 | - | 7 | 895 | ||
| BC096547.1 | - | 11 | 1410 | 30360184 | ||
| KC211186.1 | - | 12 | 1448 | |||