siRNA Details
| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145611.4 | NP_663586.3 | 11 | 2582 | ||
| GenBank mRNA | AK031324.1 | - | 11 | 2575 | ||
| AK045492.1 | - | 11 | 2554 | |||
| AK049288.1 | - | 11 | 2583 | |||
| BC006647.1 | - | 11 | 2570 | |||
| BC010245.1 | - | 10 | 2451 | 3710765 | ||

Kank2
Genomic Map