siRNA Details
| Gene Symbol: Msantd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146222.2 | NP_666334.2 | 2, 3 | 772 | ||
| GenBank mRNA | AK153989.1 | - | 1, 2 | 125 | ||
| AK196961.1 | - | 2, 3 | 208 | |||
| BC024479.1 | - | 1, 2 | 197 | 4977181 | ||
| BC066002.1 | - | 2, 3 | 772 | 5701381 | ||

Msantd2
Genomic Map