siRNA Details
| Gene Symbol: Gramd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172768.1 | NP_766356.1 | 6, 7 | 972 | ||
| GenBank mRNA | AK020827.1 | - | 6, 7 | 863 | ||
| AK028277.1 | - | 7, 8 | 867 | |||
| AK033156.1 | - | 6, 7 | 972 | |||
| AK122465.1 | - | 5, 6 | 932 | |||
| BC070451.1 | - | 5, 6 | 806 | 30616509 | ||

Gramd1b
Genomic Map