siRNA Details
| Gene Symbol: Tmem136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034863.3 | NP_001030035.1 | 3 | 545 | ||
| GenBank mRNA | AK128910.1 | - | 3 | 594 | ||
| AK158698.1 | - | 3 | 546 | |||
| AK214821.1 | - | 1 | 12 | |||
| BC139046.1 | - | 3 | 532 | 8862068 | ||
| BC139047.1 | - | 3 | 532 | 8862069 | ||

Tmem136
Genomic Map