siRNA Details
| Gene Symbol: Ttc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172770.3 | NP_766358.1 | 15, 16 | 1405 | ||
| GenBank mRNA | AK054350.1 | - | 15, 16 | 1406 | ||
| AK198378.1 | - | 2, 3 | 152 | |||
| BC056616.1 | - | 14, 15 | 1333 | 4974294 | ||

Ttc12
Genomic Map