siRNA Details
| Gene Symbol: Hykk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177351.4 | NP_796325.2 | 3 | 952 | ||
| GenBank mRNA | AK083218.1 | - | 2 | 3546 | ||
| AK085527.1 | - | 3 | 961 | |||
| AK138851.1 | - | 3 | 938 | |||
| AK142509.1 | - | 2 | 3548 | |||
| AK162425.1 | - | 3 | 953 | |||
| AK163926.1 | - | 3 | 932 | |||
| BC038618.1 | - | 3 | 931 | 4221933 | ||

Hykk
Genomic Map