siRNA Details
Gene Symbol: Lingo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181074.4 | NP_851419.1 | 4 | 916 | ||
GenBank mRNA | AF373780.1 | - | 1 | 468 | ||
AK027262.2 | - | 2 | 539 | |||
AK034084.1 | - | 2 | 1016 | |||
BC052384.1 | - | 4 | 916 | 5703857 | ||
BC065696.1 | - | 4 | 861 | 5685897 |
