siRNA Details
| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031141.1 | - | 12 | 1241 | ||
| AK158516.1 | - | 10 | 1056 | |||
| AK161676.1 | - | 11 | 1184 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp3| Gene Symbol: Usp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031141.1 | - | 12 | 1241 | ||
| AK158516.1 | - | 10 | 1056 | |||
| AK161676.1 | - | 11 | 1184 | |||