siRNA Details
| Gene Symbol: Cd109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153098.3 | NP_694738.1 | 33 | 4617 | ||
| GenBank mRNA | AK029247.1 | - | 33 | 4391 | ||
| AK036799.1 | - | 33 | 4381 | |||
| AK042169.1 | - | 7 | 906 | |||
| AK161188.1 | - | 12 | 1692 | |||
| AY083458.1 | - | 33 | 4623 | |||
| BC052443.1 | - | 33 | 4385 | 6406841 | ||
| DQ832284.1 | - | 33 | 4623 | |||

Cd109
Genomic Map