siRNA Details
| Gene Symbol: Pxylp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289645.1 | NP_001276574.1 | 7 | 1654 | ||
| NM_001289646.1 | NP_001276575.1 | 7 | 1516 | |||
| NM_001289647.1 | NP_001276576.1 | 6 | 1418 | |||
| NM_153420.3 | NP_700469.1 | 7 | 1636 | |||
| GenBank mRNA | AK036973.1 | - | 7 | 1655 | ||
| AK043758.1 | - | 6 | 1374 | |||
| AK082059.1 | - | 6 | 1374 | |||
| AK083712.1 | - | 6 | 1355 | |||
| AK084605.1 | - | 6 | 1374 | |||
| AK144699.1 | - | 7 | 1476 | |||
| AK164254.1 | - | 6 | 1345 | |||
| BC023960.1 | - | 7 | 1585 | 5323284 | ||
| BC036342.1 | - | 6 | 1363 | 4948271 | ||

Pxylp1
Genomic Map