siRNA Details
| Gene Symbol: Pxylp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289645.1 | NP_001276574.1 | 5 | 740 | ||
| NM_001289646.1 | NP_001276575.1 | 5 | 602 | |||
| NM_001289647.1 | NP_001276576.1 | 4 | 504 | |||
| NM_153420.3 | NP_700469.1 | 5 | 722 | |||
| GenBank mRNA | AK036973.1 | - | 5 | 741 | ||
| AK043758.1 | - | 4 | 460 | |||
| AK082059.1 | - | 4 | 460 | |||
| AK083712.1 | - | 4 | 441 | |||
| AK084605.1 | - | 4 | 460 | |||
| AK144699.1 | - | 5 | 562 | |||
| AK164254.1 | - | 4 | 431 | |||
| BC023960.1 | - | 5 | 671 | 5323284 | ||
| BC036342.1 | - | 4 | 449 | 4948271 | ||

Pxylp1
Genomic Map