siRNA Details
| Gene Symbol: Parp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145619.2 | NP_663594.2 | 9 | 1377 | ||
| GenBank mRNA | AF368233.1 | - | 9 | 1377 | ||
| AK044223.1 | - | 9 | 1370 | |||
| AK088125.1 | - | 9 | 1383 | |||
| AK145259.1 | - | 9 | 1411 | |||
| AK147868.1 | - | 9 | 1676 | |||
| AK165070.1 | - | 7 | 1627 | |||
| AK170541.1 | - | 9 | 1321 | |||
| BC014870.1 | - | 9 | 1317 | 3602116 | ||
| BC058754.1 | - | 9 | 1297 | 6476689 | ||
| BC062204.1 | - | 9 | 1297 | |||

Parp3
Genomic Map