siRNA Details
| Gene Symbol: Camkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145621.2 | NP_663596.1 | 6 | 639 | ||
| GenBank mRNA | AK147323.1 | - | 6 | 627 | ||
| BC017634.1 | - | 6 | 639 | 4527857 | ||
| BC022169.1 | - | 5 | 848 | |||
| BC086762.1 | - | 5 | 837 | |||
| BC111033.1 | - | 6 | 634 | 5368225 | ||

Camkv
Genomic Map