siRNA Details
| Gene Symbol: Setd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081340.2 | NP_001074809.2 | 12 | 5641 | ||
| GenBank mRNA | AK046730.1 | - | 1 | 146 | ||
| AK173246.1 | - | 1 | 62 | |||
| BC059049.1 | - | 1 | 164 | |||
| CX564178.1 | - | 4 | 547 | |||

Setd2
Genomic Map