siRNA Details
| Gene Symbol: Rtp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ428064.1 | - | 2 | 653 | ||
| AY562227.1 | - | 2 | 551 | |||
| BC112903.1 | - | 2 | 653 | 4196964 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtp3| Gene Symbol: Rtp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ428064.1 | - | 2 | 653 | ||
| AY562227.1 | - | 2 | 551 | |||
| BC112903.1 | - | 2 | 653 | 4196964 | ||