siRNA Details
| Gene Symbol: Trim43b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170884.1 | NP_001164355.1 | 7 | 1371 | ||
| Gene Symbol: Trim43c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177858.1 | NP_001171329.1 | 7 | 1371 | ||

Trim43b
Genomic Map