siRNA Details
| Gene Symbol: Mia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177389.3 | NP_796363.2 | 27 | 5735 | ||
| GenBank mRNA | AK078951.1 | - | 5 | 526 | ||
| AK220252.1 | - | 13 | 1303 | |||
| BC012393.1 | - | 1 | 91 | |||
| BC038889.1 | - | 3 | 436 | |||
| BC125472.1 | - | 13 | 1296 | |||

Mia3
Genomic Map