siRNA Details
| Gene Symbol: Usp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145628.4 | NP_663603.3 | 10 | 1391 | ||
| GenBank mRNA | AK031767.1 | - | 10 | 1370 | ||
| AK049053.1 | - | 10 | 1805 | |||
| AK169116.1 | - | 2 | 214 | |||
| AK220478.1 | - | 9 | 1104 | |||
| BC005470.1 | - | 8 | 960 | 3496607 | ||

Usp11
Genomic Map