siRNA Details
| Gene Symbol: Olfr1321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207631.1 | NP_997514.1 | 1 | 475 | ||
| GenBank mRNA | BC119046.1 | - | 1 | 542 | 8733795 | |
| BC119381.1 | - | 1 | 542 | 8734130 | ||
More Information
Set Membership: |
|

Olfr1321
Genomic Map