siRNA Details
| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172779.4 | NP_766367.1 | 10 | 1572 | ||
| GenBank mRNA | AK031945.1 | - | 10 | 1564 | ||
| AK083942.1 | - | 10 | 1534 | |||
| AK160627.1 | - | 10 | 1532 | |||
| BC051161.1 | - | 10 | 1554 | 6466142 | ||

Ddx26b
Genomic Map