siRNA Details
| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172779.4 | NP_766367.1 | 14, 15 | 2382 | ||
| GenBank mRNA | AK083942.1 | - | 14, 15 | 2344 | ||
| AK144026.1 | - | 4, 5 | 2227 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx26b| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172779.4 | NP_766367.1 | 14, 15 | 2382 | ||
| GenBank mRNA | AK083942.1 | - | 14, 15 | 2344 | ||
| AK144026.1 | - | 4, 5 | 2227 | |||