siRNA Details
| Gene Symbol: Magea10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085506.1 | NP_001078975.1 | 4 | 368 | ||
| GenBank mRNA | AK136064.1 | - | 4 | 369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magea10| Gene Symbol: Magea10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085506.1 | NP_001078975.1 | 4 | 368 | ||
| GenBank mRNA | AK136064.1 | - | 4 | 369 | ||