siRNA Details
Gene Symbol: Stard8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199018.2 | NP_950183.1 | 8 | 2214 | ||
GenBank mRNA | AK133927.1 | - | 8 | 2162 | ||
AK149830.1 | - | 8 | 2145 | |||
AK150259.1 | - | 8 | 2145 | |||
BC034186.1 | - | 4 | 1336 | |||
BC062944.1 | - | 8 | 2129 | 30357177 | ||
BC078461.1 | - | 7 | 1977 |

More Information
Panther Classification:
Molecular Function -



Gene Ontology Categories:
