siRNA Details
| Gene Symbol: Klhl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290477.1 | NP_001277406.1 | 2 | 587 | ||
| NM_001290478.1 | NP_001277407.1 | 2 | 587 | |||
| NM_172781.2 | NP_766369.2 | 2 | 587 | |||
| GenBank mRNA | AK044930.1 | - | 2 | 318 | ||
| AK081448.1 | - | 2 | 588 | |||
| AK081937.1 | - | 2 | 400 | |||
| AK085099.1 | - | 2 | 588 | |||
| AK161786.1 | - | 2 | 481 | |||
| BC057137.1 | - | 2 | 318 | |||
| BC099954.1 | - | 2 | 414 | 30081872 | ||
| BC141251.1 | - | 2 | 330 | 9055863 | ||

Klhl4
Genomic Map