siRNA Details
| Gene Symbol: Lhfpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178358.3 | NP_848135.1 | 3 | 769 | ||
| GenBank mRNA | AK142526.1 | - | 3 | 826 | ||
| AY253666.1 | - | 3 | 825 | |||
| BC051434.1 | - | 3 | 769 | 6389932 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhfpl1| Gene Symbol: Lhfpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178358.3 | NP_848135.1 | 3 | 769 | ||
| GenBank mRNA | AK142526.1 | - | 3 | 826 | ||
| AY253666.1 | - | 3 | 825 | |||
| BC051434.1 | - | 3 | 769 | 6389932 | ||