siRNA Details
Gene Symbol: Adgrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079847.2 | NP_001073316.1 | 24 | 2377 | ||
NM_001079848.2 | NP_001073317.1 | 23 | 2347 | |||
NM_001079857.2 | NP_001073326.1 | 24 | 2386 | |||
NM_001290445.1 | NP_001277374.1 | 24 | 2380 | |||
NM_001290446.1 | NP_001277375.1 | 25 | 2419 | |||
NM_178712.4 | NP_848827.1 | 25 | 2428 | |||
GenBank mRNA | AF538952.1 | - | 25 | 2393 | ||
AF538955.1 | - | 24 | 2345 | |||
AF538956.1 | - | 25 | 2384 | |||
AF538957.1 | - | 23 | 2312 | |||
AK041291.1 | - | 26 | 2617 | |||
AK046871.1 | - | 24 | 2353 | |||
BC115874.1 | - | 23 | 2322 | 40086356 | ||
BC116644.1 | - | 24 | 2373 | 40086357 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
