siRNA Details
| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146239.2 | NP_666351.2 | 15 | 1755 | ||
| GenBank mRNA | BC031778.1 | - | 1302 | 4501115 | ||
| BC049904.1 | - | 1302 | 4501115 | |||
| BC064815.1 | - | 15 | 1740 | 6516981 | ||
| BQ442862.1 | - | 5 | 339 | |||

Cdk17
Genomic Map