siRNA Details
| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146241.2 | NP_666353.1 | 16 | 2760 | ||
| GenBank mRNA | AK029833.1 | - | 2090 | |||
| AK034093.1 | - | 16 | 2139 | |||
| BC032288.1 | - | 16 | 2765 | 5368580 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trhde| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146241.2 | NP_666353.1 | 16 | 2760 | ||
| GenBank mRNA | AK029833.1 | - | 2090 | |||
| AK034093.1 | - | 16 | 2139 | |||
| BC032288.1 | - | 16 | 2765 | 5368580 | ||