siRNA Details
| Gene Symbol: Ankrd52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172790.2 | NP_766378.1 | 11, 12 | 1292 | ||
| GenBank mRNA | AK032429.1 | - | 11, 12 | 1295 | ||
| AK035533.1 | - | 11, 12 | 1280 | |||
| AK090124.1 | - | 11, 12 | 1292 | |||
| AK144017.1 | - | 11, 12 | 1302 | |||
| BC117907.1 | - | 11, 12 | 1243 | 40097189 | ||
| BC117908.1 | - | 11, 12 | 1243 | 40097194 | ||

Ankrd52
Genomic Map